Skip to Content
Merck
All Photos(1)

Key Documents

330728P

Avanti

C13 Galactosyl(β) Ceramide-d7 (d18:1-D7/13:0)

Avanti Research - A Croda Brand

Synonym(s):

D-galactosyl-β-1,1′-N-tridecanoyl-D-erythro-sphingosine-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H64D7NO8
CAS Number:
Molecular Weight:
665.00
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (330728P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(O)[C@@]([H])(NC(CCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H](O)[C@H]1CO

InChI

1S/C37H71NO8/c1-3-5-7-9-11-13-15-16-17-18-20-22-24-26-31(40)30(29-45-37-36(44)35(43)34(42)32(28-39)46-37)38-33(41)27-25-23-21-19-14-12-10-8-6-4-2/h24,26,30-32,34-37,39-40,42-44H,3-23,25,27-29H2,1-2H3,(H,38,41)/b26-24+/t30-,31+,32+,34-,35-,36+,37+/m0/s1/i1

InChI key

KJMFYHPLIHBVNV-SXOXVQABSA-N

General description

C13 Galactosyl(β) Ceramide-d7 (d18:1-D7/13:0) is deuterated derivative of C13 Galactosyl(β) Ceramide. β-Galactosyl ceramide (β −GalCer) is one of the uncomplicated glycosphingolipid. It contains a lipophilic sphingosine and a hydrophilic galactose moiety, which is attached to sphingosine via an ether linkage. It is localized in the myelin sheath of the peripheral and central nervous system.

Biochem/physiol Actions

β-Galactosyl ceramide (β −GalCer) plays a role in regulating the activity of β-glucocerebrosidase (β-GlcCer′ase). It is also a receptor for various bacterial and viral toxins.

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis, and evaluation of beta-galactosylceramide mimics promoting beta-glucocerebrosidase activity in keratinocytes
Fukunaga K, et al.
Bioorganic & Medicinal Chemistry Letters, 13(5), 813-815 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service