Key Documents
Z763284
Sigma-Aldrich
Aldrich® Sure/Seal joint adapter
with PTFE-faced EPDM rubber septum, PTFE body, joint: 14/20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
material
PTFE body
Joint
joint: 14/20
Related Categories
General description
This adapter enables repeated cannulation into any jointed flask or tube using the Sure/Seal system of PTFE-faced EPDM rubber septum mounted in an all-PTFE body adapter. Complete adapter includes PTFE body, one septum, one polypropylene overcap with hole and plastic joint clip.
  • Precision machined PTFE body with 14/20 or 24/40 joint and cap threads
  • Replaceable PTFE-faced EPDM rubber septum has excellent resistance to solvents and reseals when punctured numerous times. Use with non-coring point needles to extend septum life
  • Polypropylene over cap with hole compresses septum into adapter body to maintain seal
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)