Z742876
Kern PNJ series precision balances
Kern PNJ 600-3M verified 965-201, weighing capacity 620 g, accuracy: 0.001 g, precision: ±0.004 g g, (Verification certificate included)
Synonym(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
About This Item
Recommended Products
AC/DC input
220 - 240 V AC, universal plug set
packaging
pack of 1 ea
manufacturer/tradename
Kern PNJ 600-3M verified 965-201
weighing capacity measuring range
620 g, accuracy: 0.001 g, precision: ±0.004 g g
platform diam.
140 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Automatic internal adjustment, guarantees high degree of accuracy and makes the balance independent of its location of use. Ideal for mobile applications which require verification, such as ambulatory gold and jewellery purchasing
- High-quality tuning fork measuring system for steady weight values and continuous weighing
- Capacity display: A bar lights up to show how much of the weighing range is still available
- Precise counting: The automatic reference weight optimisation of reference weight gradually improves the averag piece weight value
- Compact size, practical for small spaces
- Large glass draught shield with 3 sliding doors for easy access to the items being weighed, for models with weighing plate size Ø 140 mm, weighing space WxDxH 172×171×160 mm
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service