Z699438
Swabs and SwabChecks
Coliform SwabCheck
Synonym(s):
colilform swab, swab kit, swab kits
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
Coliform SwabCheck
packaging
pkg of 25 ea
manufacturer/tradename
Maine Mfg 10498406
application(s)
environmental
food and beverages
suitability
bacteria
Looking for similar products? Visit Product Comparison Guide
General description
Coliform SwabCheck
Escherichia coli and coliforms are used traditionally as indicator organisms for fecal contamination in water and other environmental samples. Detection of these organisms usually points to poor hygiene at some stage in the production process or pollution of water at source. The presence of coliforms is indicated by a color change from red to yellow. The more rapid the color change the higher the level of coliform bacteria.
Escherichia coli and coliforms are used traditionally as indicator organisms for fecal contamination in water and other environmental samples. Detection of these organisms usually points to poor hygiene at some stage in the production process or pollution of water at source. The presence of coliforms is indicated by a color change from red to yellow. The more rapid the color change the higher the level of coliform bacteria.
The SwabCheck principle
The surface is wiped with a cellulose swab and any bacteria collected are transferred via the swab into a tube containing a special medium with an indicator dye, which is then incubated. A single bacterium is sufficient to cause a color change. This means that SwabCheck is about 1000 times more sensitive than the conventional ATP method. This accuracy is particularly important in the food industry. With this simple method, it is possible to identify microorganisms such as Listeria monocytogenes, which must not be present in any concentration in food and beverages.
Features and Benefits
Handling is easy. Open the sterile pack, remove the swab and wipe it over an area of about 10 x 10 cm. Then twist off the cap of the medium tube and insert the swab so that the cap fits tightly. Label the sample tube and incubate at the appropriate temperature.
A change in color indicates the presence of the microorganism in question. The quicker the color change occurs, the higher the bioburden. If no color change has been observed after the maximum incubation period has elapsed, then the corresponding microorganism is not present.GVS offers SwabCheck in packs of 25 pieces. Shelf-life of 12 months.
The surface is wiped with a cellulose swab and any bacteria collected are transferred via the swab into a tube containing a special medium with an indicator dye, which is then incubated. A single bacterium is sufficient to cause a color change. This means that SwabCheck is about 1000 times more sensitive than the conventional ATP method. This accuracy is particularly important in the food industry. With this simple method, it is possible to identify microorganisms such as Listeria monocytogenes, which must not be present in any concentration in food and beverages.
Features and Benefits
- The right test for each type of contamination
- Qualitative and semi-quantitative hygiene control
- Sterile packed and ready-for-use
- Easy to handle
- Rapid results
- Long shelf-life
- SwabCheck use
Handling is easy. Open the sterile pack, remove the swab and wipe it over an area of about 10 x 10 cm. Then twist off the cap of the medium tube and insert the swab so that the cap fits tightly. Label the sample tube and incubate at the appropriate temperature.
A change in color indicates the presence of the microorganism in question. The quicker the color change occurs, the higher the bioburden. If no color change has been observed after the maximum incubation period has elapsed, then the corresponding microorganism is not present.GVS offers SwabCheck in packs of 25 pieces. Shelf-life of 12 months.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service