Key Documents
Z679004
RF1000 pipette filler filters
pkg of 5 ea
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
packaging
pkg of 5 ea
manufacturer/tradename
Heathrow Scientific HS5000RFFIL
General description
Replacement hydrophobic 0.45 μm filters for RF1000 pipette filler.
Legal Information
RF1000 is a trademark of Heathrow Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)