Skip to Content
Merck
All Photos(1)

Key Documents

Z648310

Sigma-Aldrich

Air-driven turbine magnetic stirrer

for stirring vessels up to 1 L, pressure 3-8 psi

Synonym(s):

air driven stirrer, magnetic mixer, mixer stirrer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103806
eCl@ss:
32100100
NACRES:
NB.72

feature

for stirring vessels up to 1 L

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F37002-0000

parameter

3-8 psi pressure

General description

Users will appreciate this clever and effective air powered magnetic stirrer for it′s ease of use and low cost. A low pressure air supply is all that is needed to power this magnetic stirrer, ideal for environments where sparking hazards from electrical sources must be eliminated. This compact stirrer operates using only 3 to 8 psi (21 - 55 kPa) air pressure and is ideal for stirring vessels up to 1 L. Tubing is provided to connect air supply making this ready to use.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service