Key Documents
Z645796
Chemical spill socks
Synonym(s):
chemical cleanup, spill cleanup, spill safety
About This Item
UNSPSC Code:
47131905
NACRES:
NB.35
manufacturer/tradename
Clear-spill CS12
availability
not available in USA and Canada, ROW
General description
For the containment and absorption of hazardous and unknown liquid spills. Each sock absorbs 3 Litres.
Choose from one of the most recent versions:
Certificates of Analysis (COA)