Z567868
Aldrich® Erlenmeyer flask with ST joint
capacity 1,000 mL, joint: ST/NS 24/40
Synonym(s):
conical flasks, glass flasks, lab flask, laboratory flask
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
flat bottom
Joint
joint neck
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
without cap
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
1,000 mL
Other Notes
Browse all Erlenmeyer flasks
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)