Key Documents
Z566128
Sigma-Aldrich
Micromatic Macrovalve flushing adapter for returnable containers
allows nitrogen to be flushed through the Coupler and solvent transfer line
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
General description
This adapter allows nitrogen to be flushed through the Micromatic MacroValve Coupler and solvent transfer line when they are disconnected from the Pure-Pac® II containers.
Legal Information
Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)