Z556173
Aldrich® Soxhlet extraction apparatus
extractor capacity 85 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
extractor capacity
85 mL
flask capacity
250 mL
thimble I.D. × H
33 mm × 80 mm
Looking for similar products? Visit Product Comparison Guide
General description
Apparatus includes extractor body, condenser, and flask.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service