Z547085
Aldrich® jacketed Buchner funnel
funnel capacity 600 mL, disc diam. 90 mm, porosity 25-50 μm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
disc diam.
90 mm
funnel capacity
600 mL
stem diam.
22 mm
pore size
25-50 μm porosity
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service