Z529516
Coors™ porcelain mortar
capacity 4000 mL, top O.D. × H 283 mm × 150 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
porcelain
manufacturer/tradename
Coorstek 66331
capacity
4000 mL
pestle L
280 mm
top O.D. × H
283 mm × 150 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Mortar with lip; glazed outside, except for bottom. Order pestle separately.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service