Key Documents
Z515868
Sigma-Aldrich
Aldrich® solvent storage flask
capacity 25 mL, joint: ST/NS 14/20
Synonym(s):
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom
single-neck
Joint
joint: ST/NS14/20
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
25 mL
General description
With ground-glass joint and glass stopcock. Use septum Z565695 or Z565679 (see Septa Section).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)