Key Documents
Z420581
Sigma-Aldrich
Perforated filter plate for Table-Top Buchner funnels
for Z420557
About This Item
UNSPSC Code:
41104925
NACRES:
NB.43
material
high-density polyethylene
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H91463-1914
filter diam.
36 in.
color
none
Related Categories
General description
Filter large volumes quickly with a polyethylene plastic table-top buchner funnel using your choice of perforated filter plate. Plates are made of 4.8mm (³/₁₆") thick HDPE with 4.8mm (³/₁₆") perforations on 11mm (⁷/₁₆") centers.
Features and Benefits
  • Removable plate for use in precious metals recovery
  • Temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF)
Choose from one of the most recent versions:
Certificates of Analysis (COA)