Z403628
IKA® disperser clamps
RH 3 Strap clamp for dispersers, 1/cs
Synonym(s):
homogenizer clamps, tissue grinder clamps
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1
manufacturer/tradename
IKA 0003008600
General description
For securing vessels against walls or for synchronizing rotation during stirring or dispersing
- For stand diameter: 9-16mm
- For vessel diameter: 40-300mm
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)