Key Documents
Z303011
Quickfit® pear-shaped flasks
capacity 100 mL, joint: ST/NS 14/23
Synonym(s):
FP100/1
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
pear-shaped flask
single-neck
Joint
joint: ST/NS14/23
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Quickfit FP100/1
availability
not available in USA and Canada
capacity
100 mL
General description
Single neck with ground-glass joint
Features and Benefits
• Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)• Ideal for micro and semi-micro distillations.• Reduced losses from thermal decomposition• Minimum hold-up
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)