Z292001
Headspace vial, clear glass, beveled top, flat bottom
volume 6 mL, O.D. × L × I.D. 22 mm × 38 mm × 12.5 mm, for use with PerkinElmer®, pkg of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear glass vial
feature
closure type crimp top vial
closure type crimp top vial
packaging
pkg of 100 ea
O.D. × L × I.D.
22 mm × 38 mm × 12.5 mm
volume
6 mL
fitting
thread for 20 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
PerkinElmer is a registered trademark of PerkinElmer Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service