Z273988
FRIO-Temp® bottle thermometer
incubator, temperature 20-50 °C
Synonym(s):
incubator temperature measurement, incubator thermometer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
incubator
feature
subdivision 0.5 °C
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B61004-0500
temperature measuring range
20-50 °C
L
115 mm
Looking for similar products? Visit Product Comparison Guide
General description
NIST traceable Enviro-Safe thermometer with PTFE-coating in an unbreakable plastic bottle, containing clear, Enviro-Safe liquid. The enclosed thermometer accurately records temperature when equipment door is open. Bottles have both Velcro and magnetic attachments for easy installation.
Application
FRIO-Temp® bottle thermometer has been used to record the sample temperature in the incubator
Features and Benefits
FRIO-Temp® bottle thermometer have the following benefits:
- The thermometer reads accurate temperature without sample contamination as the filled bottle (included) mimics the sample.
- For best results insert the thermometer into the bottle and place it near the sample.
- The thermometer consists of organic liquid or Enviro-Safe® liquid.
- The thermometer has lead free glass and permanently fused markings.
Legal Information
Enviro-Safe is a registered trademark of H-B Instrument Co.
FRIO-Temp is a registered trademark of SP Scienceware, a division of SP
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service