Key Documents
Z266094
Supelco
TLC plate storage racks
Rack for 20 cm × 20 cm plates
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
material
stainless steel (20 gauge)
technique(s)
thin layer chromatography (TLC): suitable
General description
TLC plate storage racks is a rack of convenient size, which is used to hold the prepared plates during the process of drying and transportation. It may be used to store the agar plates used in agarose gel electrophoresis.
Used to carry and store TLC plates without damage to media. Racked plates may be air or oven dried. Holds up to 10 standard plates in either a vertical or horizontal position. Racks are stainless steel with a carrying handle.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Laboratory diagnosis of specific organ diseases by means of combined serum isoenzyme patterns.
Nerenberg TS and Pogojeff G
American Journal of Clinical Pathology, 51(4), 429-439 (1969)
Food Chemicals Codex (2003)