Z260231
FORTUNA® Optifix® Safety bottle-top dispenser
Safety model, volume 10-50 mL
All Photos(1)
About This Item
Recommended Products
feature
subdivision 1.0 mL
manufacturer/tradename
Fortuna Graf 101.107-48
volume measuring range
10-50 mL
volume
10-50 mL
fitting
GL 40
GL 45
Related Categories
General description
For dispensing strong acids and aggressive chemicals
Features:
Features:
- PTFE encased dispensing piston with sealing lip at its end
- FIX adapter made of PTFE
- Valve block made of PTFE, no metal springs in the valve - no corrosion
- Visible dispensing process by the use of a dispensing cylinder made of glass, coated with plastic-protection against breakage
- Ergonmic finger protection cap
- Fast and easy volume setting with additional fine adjustment
Legal Information
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)