Z243914
Aldrich® TLC developing tank - cylindrical
glass tank, O.D. × H 6.5 cm × 21.0 cm
Synonym(s):
Cylindrical TLC tank
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
thin layer chromatography (TLC): suitable
O.D. × H
6.5 cm × 21.0 cm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Can be used as a staining chamber. Made of glass with lid. Compact size offers good plate visibility and minimal solvent use.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service