Z225118
Large lab spoon
stainless steel, L 12 in.
Synonym(s):
sampling spoon
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F36711-0012
L
12 in.
General description
Spoon (1.5 in. wide) with a flat spatula on one end (1.2 in. wide).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service