Skip to Content
Merck
All Photos(3)

Key Documents

Z193208

Micro spatulas

Curved at one end with knob at the other end, L 5 1/12 in. (140 mm)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122403
NACRES:
NB.35

material

stainless steel

packaging

pkg of 2 ea

L

5 1/12 in. (140 mm)

Looking for similar products? Visit Product Comparison Guide

General description

Curved, square-cut, sharpened tip (3 mm wide) at one end, with small knob at the other end.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service