Z174777
Aldrich® jacketed chromatography column
column capacity 400 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
for chromatography
technique(s)
LPLC: suitable
column capacity
400 mL
reservoir capacity
500 mL
weight
250 gMax. packing
Looking for similar products? Visit Product Comparison Guide
General description
24/40 joint, gravity fed, PTFE stopcock. A slight nitrogen pressure may be applied using an inlet adapter. Recommended packing: silica gel 923 (Aldrich Cat. No. 214477). For low-temperature chromatography, circulate cooling liquid through outer jacket.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service