Key Documents
Z173169
Sigma-Aldrich
Aldrich® Schlenk filter tube
capacity 75 mL, porosity 25-50 μm, male Joints: ST/NS 14/20 (2), female Joints: ST/NS 14/20 (2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
colorless tube
round bottom
Joint
female Joints: ST/NS14/20 (2)
male Joints: ST/NS14/20 (2)
sterility
non-sterile
feature
graduations: no
capacity
75 mL
pore size
25-50 μm porosity
binding type
non-treated surface
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)