Key Documents
Z170119
Sigma-Aldrich
Scienceware® glove box bare-hand port
iris port
Synonym(s):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
UNSPSC Code:
42132201
NACRES:
NB.35
label
E
packaging
pack of 2 ea
manufacturer/tradename
Bel-Art H50025-0020
General description
The Scienceware glove box bare-hand port is for use when operations with gloves are difficult. Allows access to any glove box with a 73/8 in. I.D. port. Constructed of seven layers, 20 points of soft conforming silicone-latex rubber for an excellent seal. The bare-hand port is used with the Scienceware glove box, Z169943.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)