Z154482
Ace polished shaft
diam. × L 10 mm × 580 mm, with button: no
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
glass
feature
for flask size 0.5 ‑ 12 L
manufacturer/tradename
Ace Glass 807523
diam. × L
10 mm × 580 mm
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service