Key Documents
Z136115
Scienceware® vernier caliper
Direct-reading type
About This Item
UNSPSC Code:
41111700
NACRES:
NB.79
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H13415-0000
General description
Metric and English scales with 0.1 mm and 0.001 in. subdivisions.
Metric/Imperial. Molded in light, corrosion-resistant plastic. 6 in. or 150 mm measuring capacity; parallel jaws are 1.57 in. (40 mm) deep; reading scales create no parallax. Nonconductive, fine-adjustment thumb roller, two-direction lock, inside-outside jaws, round-rod depth gauge. Upper scale reads in mm, lower scale in inches.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)