Key Documents
Z131008
Sigma-Aldrich
Nalgene® beaker with handle
capacity 3,000 mL
Synonym(s):
HDPE beaker, heavy wall beaker
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
high-density polyethylene
feature
graduations
packaging
pack of 1 ea
manufacturer/tradename
Nalgene 1220-2500
capacity
3,000 mL
General description
Heavy-wall, HDPE, rectangular handle. Graduated in oz and mL. Suitable for handling acids and alkalis.
Other Notes
Browse all plastic beakers
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)