Z118125
Hamilton® needles for RN syringes
volume 2.5-100 μL, needle size 22 ga, bevel tip, needle L 101.5 mm (4 in.)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
stainless steel needle (point style 4)
needle size
22 ga
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 7804-01
needle L
101.5 mm (4 in.)
volume
2.5-100 μL
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
22° bevel
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service