Skip to Content
Merck
All Photos(1)

Key Documents

WHA67041500

Whatman® carbon/HEPA filter capsule

Carbon Cap 150

Synonym(s):

Whatman filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161807
NACRES:
NB.24

material

glass fiber membrane (With activated carbon bed)
polypropylene housing

sterility

non-sterile

packaging

pack of 1 ea

manufacturer/tradename

Whatman 6704-1500

diam.

150 mm

pore size

>0.3 μm particle size

fitting

male inlet
male outlet (Sliptip)

Looking for similar products? Visit Product Comparison Guide

General description

Carbon Cap capsule filters are filled with activated carbon and include a pleated glass fiber filter. They are suitable for adsorption of organic vapors from air, and for removing color, organics, and chlorine from water.

Carbon cap for organic and chlorine filtration

Large volume chlorine and organic filtration are facilitated by activated carbon, which adsorbs these molecules. Carbon Cap is a filter filled with high-purity, high-efficiency, acid-washed, activated carbon and a High Efficiency Particulate Air (HEPA) filter. It is made specifically for adsorption of organic vapors, chlorine, and decolorizing liquids.

Application

  • Water, chemical, and reagent purification
  • Removes noxious odors, oil mists, and contaminants
  • Compressed air lines and vacuum pumpsInstrument outlet exhausts
  • Eliminates a potential health hazard from the workplace

Features and Benefits

  • Activated carbon acts as an adsorption media for color, organics, and chlorine
  • Pleated glass microfiber filter structure increases surface area, resulting in high efficiency
  • Retains 99.97% of particles greater than 0.3 µm

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service