Key Documents
WHA10477601
Whatman® Vacuum filtration apparatus accessories
WT 100 Witt′s flask, with tubing nozzle, capacity
Synonym(s):
Z751960
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
packaging
pk of 1
manufacturer/tradename
Whatman 10477601
Whatman Article No. 28415990 (US reference)
capacity
General description
Borosilicate glass bottle for filtrate collection in an inserted container.
Features and Benefits
  • For filtrate collection in an inserted container.
  • The bottle is made of borosilicate glass.
  • Has a replaceable round lid and side-mounted tubing nozzle for vacuum tubing 8 mm (inside diameter).
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)