WHA10347893
Whatman® qualitative filter paper, Grade 2122
W × L 100 mm × 100 mm
Synonym(s):
Whatman filter, Whatman paper, Z696358, paper filter
About This Item
Recommended Products
material
paper filters (parchment)
sheets
feature
Grade 2122
packaging
pkg of 500 ea
manufacturer/tradename
Whatman 10347893
Whatman Article No. 28414668 (US reference)
W × L
100 mm × 100 mm
basis weight
40 g/m2
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Smooth surface allows samples to slide off with ease and not be retained on paper.
- Shiny pergamyne paper allows visual determination that all sample has been removed.
- Extremely thin (0.03 mm) and lightweight (40 g/m²) for safe and secure sample transport.
- No glue or additives (residue-free).
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service