Skip to Content
Merck
All Photos(1)

Key Documents

W400605

Sigma-Aldrich

L-Monomenthyl glutarate

≥72%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O4
CAS Number:
Molecular Weight:
270.36
FEMA Number:
4006
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.929
NACRES:
NA.21
Organoleptic:
minty
grade:
Fragrance grade
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

grade

Fragrance grade

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

Assay

≥72%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

minty

SMILES string

O([C@H]1[C@@H](CC[C@H](C1)C)C(C)C)C(=O)CCCC(=O)O

InChI

1S/C15H26O4/c1-10(2)12-8-7-11(3)9-13(12)19-15(18)6-4-5-14(16)17/h10-13H,4-9H2,1-3H3,(H,16,17)/t11-,12+,13-/m1/s1

InChI key

CTMTYSVTTGVYAW-FRRDWIJNSA-N

Related Categories

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service