W362320
5-Ethyl-4-hydroxy-2-methyl-3(2H)-furanone
≥96%, natural (US), FG
Synonym(s):
Ethyl furanone, Homofuraneol
About This Item
Recommended Products
biological source
botanical
Quality Level
grade
FG
Fragrance grade
Halal
Kosher
natural (US)
Agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.510
Assay
≥96%
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
berry; wine-like
storage temp.
2-8°C
InChI
1S/C7H10O3/c1-3-5-7(9)6(8)4(2)10-5/h4,9H,3H2,1-2H3
InChI key
QJYOEDXNPLUUAR-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
183.2 °F - closed cup
Flash Point(C)
84 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service