Skip to Content
Merck
All Photos(1)

Key Documents

W353977

Sigma-Aldrich

Ocimene

mixture of isomers, stabilized, ≥90%

Synonym(s):

3,7-Dimethyl-1,3,6-octatrien, 3,7-Dimethyl-1,3,6-octatriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
FEMA Number:
3539
Beilstein:
1736172
MDL number:
UNSPSC Code:
41116107
Flavis number:
1.018
NACRES:
NA.21

Agency

meets purity specifications of JECFA

Quality Level

reg. compliance

FDA 21 CFR 117

Assay

≥90%

contains

α-Tocopherol

refractive index

n20/D 1.485

bp

65-66 °C/13 mmHg

density

0.818 g/mL at 20 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; woody; tropical

SMILES string

[H]C(CC([H])=C(C)C=C)=C(C)C

InChI

1S/C10H16/c1-5-10(4)8-6-7-9(2)3/h5,7-8H,1,6H2,2-4H3

InChI key

IHPKGUQCSIINRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Self-heat. 2 - Skin Irrit. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service