Skip to Content
Merck
All Photos(1)

Key Documents

W327312

Sigma-Aldrich

2,6-Dimethylpyrazine

natural (US)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
FEMA Number:
3273
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
14.021
NACRES:
NA.21
grade:
Fragrance grade
Halal
Kosher
natural (US)
food allergen:
no known allergens

grade

Fragrance grade
Halal
Kosher
natural (US)

Quality Level

300
400

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

Assay

≥90%

bp

154 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coffee; meaty; nutty

SMILES string

Cc1cncc(C)n1

InChI

1S/C6H8N2/c1-5-3-7-4-6(2)8-5/h3-4H,1-2H3

InChI key

HJFZAYHYIWGLNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Pyrazines Biosynthesis by Bacillus Strains Isolated from Natto Fermented Soybean.: This study explores the biosynthesis of pyrazines, including 2,6-Dimethylpyrazine, by Bacillus strains isolated from natto fermented soybean. The research highlights the biochemical pathways and potential applications in flavor and aroma enhancement in food products, providing insights into the natural production of these compounds (Kłosowski et al., 2021).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service