Skip to Content
Merck
All Photos(1)

Key Documents

W235105

Sigma-Aldrich

Cyclohexyl butyrate

FG

Synonym(s):

Butanoic acid cyclohexyl ester, Butyric acid cyclohexyl ester, Cyclohexyl butanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2C6H11
CAS Number:
Molecular Weight:
170.25
FEMA Number:
2351
EC Number:
Council of Europe no.:
2082
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.230
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

98%

refractive index

n20/D 1.4425 (lit.)

bp

202-203 °C/700 mmHg (lit.)

density

0.943 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; pineapple

SMILES string

CCCC(=O)OC1CCCCC1

InChI

1S/C10H18O2/c1-2-6-10(11)12-9-7-4-3-5-8-9/h9H,2-8H2,1H3

InChI key

VZHUBBUZNIULNM-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service