Skip to Content
Merck
All Photos(1)

Key Documents

TEM-LC200CU25

Sigma-Aldrich

Lacey Carbon Supported Copper Grids

size 200 mesh, size 50 nm , hole size, box of 25 ×

Synonym(s):

Lacey/Carbon Film, 200 Mesh, Copper, 25pk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
MA.24

packaging

box of 25 ×

size

200 mesh
50 nm , hole size

Looking for similar products? Visit Product Comparison Guide

General description

Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.

Application

  • Optimizing "self-wicking" nanowire grids.: This study focuses on the development and optimization of self-wicking nanowire grids, emphasizing the significance of lacey carbon supported copper grids in enhancing the performance of nanowire-based applications. The research provides insights into improving grid design for better electron microscopy imaging and potential applications in various industrial sectors (Wei et al., 2018).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service