TEM-HC200CU100
Holey Carbon Supported Copper Grids
size 200 mesh, size 100 nm , Hole, box of 25
Synonym(s):
Holey Carbon - Cu, 200 mesh, 100 micron
All Photos(1)
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Recommended Products
material
carbon film (holey)
copper grid
packaging
box of 25
size
100 nm , Hole
200 mesh
Related Categories
General description
Holey carbon film supported copper TEM gird: hole size 100 nm on 300 mesh copper grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)