SYNF304100
Synthware™ single-neck round-bottom flask
100 mL, joint: ST/NS 24/40
Synonym(s):
Synthware F304100, glass round flask
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Recommended Products
material
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F304100
capacity
100 mL
100 mL
Related Categories
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)