SYNF161450
Synthware™ two neck reaction flask with glass stopcock
capacity 50 mL, joint: ST/NS 14/20
Synonym(s):
Synthware F161450, flask, reaction flask, two-neck flask
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Recommended Products
material
borosilicate glass flask
round bottom
two-neck
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F161450
capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
With a 2 mm glass stopcock having a constant pressure retention device.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service