SYNE112585X
Synthware™ extraction thimble
O.D. 25 mm, porosity: extra coarse, H 85 mm
Synonym(s):
Synthware E112585X, glass thimble
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Synthware E112585X
H
85 mm
O.D.
25 mm
General description
All glass extraction thimble permits viewing extraction process and subsequent weighing. C-Coarse, XC-Extra Coarse. Small thimble is used with C243445 and E103424; Medium thimble is used with C244500 and E104524; Large thimble is used with C245550 and E105550.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)