Key Documents
SYNC504300
Synthware Liebig condenser with 8 mm OD removable hose connections
joint: ST/NS 24/40, jacket L 300 mm
Synonym(s):
Synthware C504300, straight condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C504300
jacket L
300 mm
Related Categories
General description
Synthware Liebig condenser features a 300 mm jacketed length for superior cooling efficiency. With standard taper outer joint at the top (24/40) and a lower inner drip tip joint. Made from durable borosilicate glass.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)