Key Documents
SYNC154200
Synthware West condenser
joint: ST/NS 24/40, jacket L 200 mm
Synonym(s):
Synthware C154200, distillation condenser, Inland Revenue condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C154200
jacket L
200 mm
Related Categories
General description
With narrow annular space for fast coolant speed, thus high cooling efficiency.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)