Key Documents
SLW1516/04D
Pyrex® Media-Lab Bottles, with cap and pouring ring, with printed trace code
capacity 100 mL
Synonym(s):
Pyrex, SciLabware 1516/04D
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
borosilicate glass bottle
polypropylene cap
round bottle
white graduation
Joint
Joints threaded neck (GL45)
sterility
non-sterile
feature
graduations
with cap
with pouring ring
with printed trace code
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1516/04D
capacity
100 mL
diam. × H
56 mm × 100 mm
General description
  • Bottles complete with polypropylene cap and drip-free pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Ideal for mixing and storage of culture medium
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
  • For use up to 140 deg. C
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)