Key Documents
SLW1000/11M
Pyrex® Griffin beakers, low form, with printed trace code
25 mL
Synonym(s):
Pyrex, SciLabware 1000/11M
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
borosilicate glass
white graduation
feature
low form
with printed trace code
manufacturer/tradename
1000/11M
SciLabware
H × diam.
50 mm × 34 mm
capacity
25 mL
General description
  • Complies with ISO 3819 and DIN 12331
  • Manufactured from Pyrex borosilicate glass
  • Excellent thermal performance
  • High resistance to chemical attack
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • White graduations and marking spot
Other Notes
Browse all glass beakers
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)