PEM3A5022
Xion PEM-Aquivion®-720
thickness 50 μm, L × W 5 cm × 5 cm
About This Item
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
L × W
5 cm × 5 cm
thickness
50 μm
greener alternative category
SMILES string
FC(CC)(OC(F)(C(F)(S(O)(O)=O)F)F)CC
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service