Recommended Products
sterility
non-sterile
form
liquid
manufacturer/tradename
Mettler-Toledo 51350058
volume
250 mL
pH
10.012
Looking for similar products? Visit Product Comparison Guide
General description
Top Precision: buffers according to NIST and DIN 19266.
The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision. These buffers also come with a detailed test certificate and are only available in individual 250 mL bottles. Calibration cannot get better than this!
The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision. These buffers also come with a detailed test certificate and are only available in individual 250 mL bottles. Calibration cannot get better than this!
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service