MT30046240
Mettler Toledo SevenExcellence™ pH/mV meter S400
S400 kit, meter only, AC/DC input 230 V AC, universal plug set
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Mettler-Toledo 30046240
mV measuring range
-2000-2000, accuracy: ±0.1
pH range
-2-20
General description
Includes meter with a pH/mV expansion unit and 2 blank expansion units
SevenExcellence™ pH/mV meter – intuitive touchscreen operation and high measurement performance combined with a complete security package for safe results and solid compliance. This single channel instrument is modularly expandable with additional measurement parameters at any time!
Professional calibration support, incl. automatic calibration with Rondolino sample changer
Large 7 inch color display and intuitive menu guidance in 10 languages
Flexible method concept for high reproducibility and security
Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
Integrated interfaces (USB, RS232 and Ethernet) for data exchange
Comprehensive service package including IQ/OQ/PQ
Included in every SevenExcellence:
uPlace™ electrode holder with a perfect vertical motion. It can be operated with one hand and used either freestanding or attached to the left or right side of the instrument.
LabX direct pH PC software for transferring real-time measurement data and archiving your results
Professional calibration support, incl. automatic calibration with Rondolino sample changer
Large 7 inch color display and intuitive menu guidance in 10 languages
Flexible method concept for high reproducibility and security
Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
Integrated interfaces (USB, RS232 and Ethernet) for data exchange
Comprehensive service package including IQ/OQ/PQ
Included in every SevenExcellence:
uPlace™ electrode holder with a perfect vertical motion. It can be operated with one hand and used either freestanding or attached to the left or right side of the instrument.
LabX direct pH PC software for transferring real-time measurement data and archiving your results
Legal Information
ISM is a registered trademark of Mettler-Toledo, Inc.
SevenExcellence is a trademark of Mettler-Toledo, Inc.
uPlace is a trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)